missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Bovine IgG F(ab')2 Secondary Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP1-73497
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Spécification
IgG F(ab')2 | |
Polyclonal | |
HRP | |
immunoglobulin heavy constant gamma 1 (G1m marker) | |
Bovine IgG F(ab')2 fragment | |
2 mg | |
Secondary | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
LYOPH | |
Western Blot 1:1000 - 1:10000, ELISA 1:40000 - 1:80000, Immunohistochemistry 1:500 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50, Knockdown Validated | |
Rabbit | |
Affinity Purified | |
RUO | |
No reaction was observed against Bovine IgG F(c). | |
Bovine | |
IgG |
For Research Use Only