Learn More
RAB10 Monoclonal Antibody (4E2), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA515670
Description
MA5-15670 targets Rab10 in indirect ELISA, IF and WB applications and shows reactivity with Human and mouse samples. The MA5-15670 immunogen is purified recombinant fragment of human Rab10 expressed in E. Coli. . MA5-15670 detects Rab10 which has a predicted molecular weight of approximately 22.5kDa.
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.
Spécification
RAB10 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P61026, P61027 | |
RAB10 | |
Purified recombinant fragment of human Rab10 expressed in E. Coli. | |
RUO | |
10890, 19325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
4E2 | |
Unconjugated | |
RAB10 | |
Ac1075; AW107754; GTP-binding protein RAB10; Rab10; RAB10, member RAS oncogene family; ras-related GTP-binding protein; ras-related protein rab10; ras-related protein Rab-10 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.