Learn More
Corning™ CoolRack™ XT CFT24, contient 24 tubes cryovial ou FACS
Contient 24 flacons cryogéniques ou tubes FACS, avec des puits de préhension pour l'ouverture/fermeture du flacon d'une seule main.
Marque:  Corning™ 432050
Code nomenclature Nacres: NB.43
Informations supplémentaires : Poids : 2.00000kg
Code produit. 15552821
  • 332.00€ / 1 pièce
Expédition estimée: 19-11-2024
Description
Corning CoolRack thermo-conductive tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
  • Ensure +/- 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible
  • "Gripping" wells for one-hand vial opening/closing
Spécification
Spécification
24 Cryogenic vial or FACS Tubes
CFT Tube Modules