missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Protirelin acetate

Code produit. 16499186
Encompass_Preferred
Click to view available options
Quantité:
100 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 16499186

Marque: MedChemExpress HYP0002A

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

MedChemExpress Protirelin Acetate is a highly conserved neuropeptide that exerts the hormonal control of thyroid-stimulating hormone (TSH) levels as well as neuromodulatory functions.

Spécification

Pureté 99.93%
CAS 120876-23-5
Contenu et stockage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
À utiliser avec (application) COVID-19-immunoregulation
Poids moléculaire 452.46 g/mol
Type de produit Peptides
Quantité 100 mg
Conditions d’expédition Room temperature
Solubilité DMSO : 250 mg/mL (552.54 mM∣Need ultrasonic) ∣H2O : 50 mg/mL (110.51 mM∣Need ultrasonic)
Synonyme Thyrotropin-releasing-hormone acetate∣TRH acetate
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.