Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
PROTEIN FREE HYBRIDOMA MED P
Manufacturer:  Gibco 12040051
85.28 EUR valable jusqu'au 2025-04-30
Utilisez le code promo "24107" pour bénéficier de cette offre.
PROTEIN FREE HYBRIDOMA MED P
Code nomenclature Nacres: NA.79
Additional Details : Poids : 0.50000kg
Catalog No. 11550416
  • 94.75€ / 500mL
Expédition estimée: 02-05-2025
pour voir le stock disponible 
Explore more special offers
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates