Learn More
Eppendorf™ Ensemble de filtres de protection
Designed for use with Eppendorf Reference™ 2, Eppendorf Research™ plus and Eppendorf Xplorer™ (plus) pipettes
Marque: Eppendorf™ 4920624001
Code nomenclature Nacres: NB.01
Informations supplémentaires : Poids : 0.01100kg
Description
- Available in a variety of colors
- Protection Filter Sets include 10 filters and 1 filter sleeve
- For fixed- and variable-volume pipettes
Spécification
Turquoise | |
10 mL | |
10 filters with 1 filter sleeve |
Protection filter | |
1 to 10 mL variable Eppendorf Reference™ 2, Eppendorf Research™ plus and Eppendorf Xplorer™ (plus) | |
1par unité |