missing translation for 'onlineSavingsMsg'
Learn More

PROTAC BRD9-binding moiety 1 hydrochloride, MedChemExpress

Product Code. 16414138
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantité:
5 mg
10 mg
25 mg
50 mg
100 mg
This item is not returnable. View return policy

Product Code. 16414138

missing translation for 'mfr': Medchem Express HY107445A

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

MedChemExpress PROTAC BRD9-binding moiety 1 hydrochloride is a compound that binds to BRD9, and used for inhibiting BRD9 activity, based on PROTAC.

PROTAC BRD9-binding moiety 1 hydrochloride is a compound that binds to BRD9, and used for inhibiting BRD9 activity, based on PROTAC.

Specifications

Nom chimique ou matériau PROTAC BRD9-binding moiety 1 hydrochloride
Couleur Light Yellow
Notes de qualité de la pureté Research
Formule moléculaire C23H26ClN3O7S2
Quantité 10 mg
Informations sur la solubilité DMSO : 20.83 mg/mL (37.46 mM; Need ultrasonic) ∣H2O : 2.5 mg/mL (4.50 mM; Need ultrasonic)
SMILES O=C(O)COC1=CC=C(C2=CN(C)C(C3=C2SC(C(NC(CC4)CCS4(=O)=O)=N)=C3)=O)C=C1OC.Cl
Poids moléculaire (g/mol) 556.05
Poids de la formule 556.05
Pourcentage de pureté 97.07%
Qualité Research
Forme physique Solid
À utiliser avec (application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.