missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PP1 alpha/PPP1A Recombinant Protein Antigen

Code produit. 18178297 Tous les produits Bio Techne Produits
Click to view available options
Quantité:
0,1 ml
Conditionnement:
0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 18178297

Marque: Novus Biologicals™ NBP238559PEP

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PPP1CA. The PP1 alpha/PPP1A Recombinant Protein Antigen is derived from E. coli. The PP1 alpha/PPP1A Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP2-38559. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

TRUSTED_SUSTAINABILITY

Spécification

Gene ID (Entrez) 5499
Espèces Human
Méthode de purification Chromatography
Pureté >80%
Concentration 0.5mg/mL
Contenu et stockage Store at -20°C. Avoid freeze-thaw cycles.
Formule PBS and 1M Urea, pH 7.4.
À utiliser avec (application) Blocking/Neutralizing, Control
Symbole de gène(s) PPP1CA
Type d’étiquette Unlabeled
Poids moléculaire 20kDa
Type de produit PP1 alpha/PPP1A
Quantité 0,1 ml
État réglementaire RUO
Source E.Coli
Réactivité spécifique This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-38559. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Immunogène QILKPADKNKGKYGQFSGLNPGGRPIT
Show More Show Less

Usage exclusivement réservé à la recherche

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.