Learn More
Thermo Scientific™ POROS™ D50 RoboColumn
RoboColumn™ chromatography products are small columns provided in 8-column strips.
Marque:  Thermo Scientific™ A30724
Code nomenclature Nacres: NB.21
Informations supplémentaires : CAS : 64-17-5 Poids : 0.13000kg
Code produit. 18406488
  • 317.00€ / 600µL
Expédition estimée: 12-08-2024
pour voir le stock disponible 
Description
Description
RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms. These particular RoboColumns contain POROS™ D50 resin, a 50-micron weak anion-exchange resin suitable for low-pressure applications, including large-scale downstream bioprocessing.
Features of the resin:
  • Higher productivity—through high throughput and high dynamic capacity associated with high performance chromatography
  • Chemical stability—allows aggressive cleaning and sanitization
  • Enhanced biomolecule access—provided via large pores (500-10,000 Å)
  • Polystyrenedivinylbenzene particles—yield a robust, easily packable matrix
  • Better separation methods in less time—the speed of high performance chromatography reduces weeks of experimentation to only a few hours, leaving more time to explore all the variables of your separation
RoboColumn is a registered trademark of ATOLL GmbH.
Spécification
Spécification
Store at 2°C to 8°C or at room temperature.
600 μL
16 to 1000 cm/hr.