missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR3H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16423-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RPC8 is DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Specific peripheric component of RNA polymerase III which synthesizes small RNAs, such as 5S rRNA and tRNAs. It is plays a key role in sensing and limiting infection by intracellular bacteria and DNA viruses. It is acting as nuclear and cytosolic DNA sensor involved in innate immune response. It can sense non-self dsDNA that serves as template for transcription into dsRNA. The non-self RNA polymerase III transcripts, such as Epstein-Barr virus-encoded RNAs (EBERs) induce type I interferon and NF-Kappa-B through the RIG-I pathway.Spécification
POLR3H | |
Polyclonal | |
Unconjugated | |
POLR3H | |
KIAA1665, POLR3H, RNA polymerase III subunit C8, RPC22.9, RPC8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171568, 300088, 78929 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9D2C6, Q9Y535 | |
POLR3H | |
POLR3H Fusion Protein Ag9624 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |