missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POLR2J Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Spécification
| Antigène | POLR2J |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Codice del prodotto | Marca | Quantité | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantité | Prezzo | Quantità e disponibilità | |||||
|
16826024
|
Proteintech
16403-1-AP-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
16816024
|
Proteintech
16403-1-AP-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Descrizione
Three genes (POLR2 J1, POLR2 J2, and POLR2 J3) form a cluster of total length of 214 530 bp in the genetic locus 7q22.1 on the long arm of chromosome 7 (contig NT_007933). The fourth gene (POLR2 J4, 31 040 bp) was localized in the cytogenetic locus 7p13 of the short arm of chromosome 7 (contig NT_007819). An analysis enabled us to refine dissimilar experimental data on the mapping of the hRPB11 subunit gene on chromosome 7. In particular, the presence of three sites of its localization according to data on hybridization with fluorescent-labeled probes (the FISH method) was explained. It was established that, upon the expression of the four human POLR2 J genes, at least 14 types of mature mRNAs encoding somewhat differing hRPB11 isoforms can be synthesized. Eleven of these mRNAs were revealed (as full-length copies or clearly identifiable fragments) in the available databases of expressed sequence tags and cDNAs. The most probable scheme of origination of the multiple genes of the POLR2 J family as a result of three consecutive segmented duplications increasing in size was proposed and substantiated. On the basis of the scheme, some assumptions on the pathways of evolution of separate human genes and the mechanisms of generation of protein diversity in higher eukaryotes were madeSpecifica
| POLR2J | |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O08740, P52435 | |
| 20022, 288588, 5439 | |
| POLR2J Fusion Protein Ag9520 | |
| Primary | |
| -20°C | |
| POLR2J |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hRPB14, POLR2J, POLR2J1, RPB11, RPB11A, RPB11m | |
| Polr2j | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto