missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PNPO Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15552-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5′-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.Spécification
PNPO | |
Polyclonal | |
Unconjugated | |
PNPO | |
PDXPO, PNPO, Pyridoxamine phosphate oxidase, Pyridoxine 5 phosphate oxidase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
103711, 55163, 64533 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O88794, Q91XF0, Q9NVS9 | |
PNPO | |
PNPO Fusion Protein Ag7914 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |