missing translation for 'onlineSavingsMsg'
Learn More

PKC zeta Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26899-1-AP-20UL

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16825235

  • 140.00€ / 20µL
Expédition estimée: 09-08-2024
pour voir le stock disponible 

Description

Description

PKC zeta is a member of the protein kinase C (PKC) family play a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function. PKCs were originally identified as serine/threonine protein kinases whose activity was dependent on calcium and phospholipids. Diacylglycerols (DAG) and tumor promoting phorbol esters bind to and activate PKC. PKCs can be subdivided into at least two major classes including conventional (c) PKC isoforms ( alpha, betaI, betaII and gamma) and novel (n) PKC isoforms ( delta, epsilon, zeta, eta and theta). Patterns of expression for each PKC isoform differ among tissues and PKC family member exhibit clear differences in their cofactor dependencies. For instance, the kinase activities of nPKC delta and epsilon are independent of Ca2+.
Spécification

Spécification

PKC zeta
Polyclonal
Unconjugated
PRKCZ
PKC ZETA, PKC2, PRKCZ, Protein kinase C zeta type, protein kinase C, zeta
Rabbit
Antigen Affinity Chromatography
RUO
18762, 5590
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q02956, Q05513
Prkcz
PKC zeta Fusion Protein Ag25391
20 μL
Primary
Human, Mouse
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions