missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC zeta Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26899-1-AP-150UL
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
PKC zeta is a member of the protein kinase C (PKC) family play a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function. PKCs were originally identified as serine/threonine protein kinases whose activity was dependent on calcium and phospholipids. Diacylglycerols (DAG) and tumor promoting phorbol esters bind to and activate PKC. PKCs can be subdivided into at least two major classes including conventional (c) PKC isoforms ( alpha, betaI, betaII and gamma) and novel (n) PKC isoforms ( delta, epsilon, zeta, eta and theta). Patterns of expression for each PKC isoform differ among tissues and PKC family member exhibit clear differences in their cofactor dependencies. For instance, the kinase activities of nPKC delta and epsilon are independent of Ca2+.Spécification
PKC zeta | |
Polyclonal | |
Unconjugated | |
PRKCZ | |
PKC ZETA, PKC2, PRKCZ, Protein kinase C zeta type, protein kinase C, zeta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18762, 5590 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q02956, Q05513 | |
Prkcz | |
PKC zeta Fusion Protein Ag25391 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |