missing translation for 'onlineSavingsMsg'
Learn More

PKC zeta Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Spécification

Antigène PKC zeta
Concentration 0.2 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16817533
Afficher les documents
Proteintech
10860-1-AP-20UL
20 μL
145.00€
20µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16807533
Afficher les documents
Proteintech
10860-1-AP-150UL
150 μL
410.00€
150µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Protein kinase C (PKC) zeta is a member of the PKC family of serine/threonine kinases which are involved in a variety of cellular processes such as proliferation, differentiation and secretion. Unlike the classical PKC isoenzymes which are calcium-dependent, PKC zeta exhibits a constitutive kinase activity which is independent of calcium, and PKC activators, phosphatidylserine and diacylglycerol. Furthermore, it is insensitive to PKC inhibitors and cannot be activated by phorbol ester. The structural and biochemical properties indicate that the zeta subspecies is related to, but distinct from other isoenzymes of PKC.
Spécification

Spécification

PKC zeta
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human
Q02956, Q05513
18762, 5590
PKC zeta Fusion Protein Ag1310
Primary
-20°C
PRKCZ
0.2 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
PKC ZETA, PKC2, PRKCZ, Protein kinase C zeta type, protein kinase C, zeta
Prkcz
IgG
Antigen Affinity Chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications