missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP5K1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12541-1-AP-20UL
This item is not returnable.
View return policy
Description
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Specifications
| PIP5K1B | |
| Polyclonal | |
| Unconjugated | |
| PIP5K1B | |
| MSS4, PIP5K1 beta, PIP5K1B, PIP5KIbeta, Protein STM 7, PtdIns(4)P 5 kinase 1 beta, STM7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18719, 309419, 8395 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14986, P70181, Q5CZZ9 | |
| Pip5k1b | |
| PIP5K1B Fusion Protein Ag3261 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction