missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-AKT2 (Ser474) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA5121255
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Positive test controls include: 293. The target is usually found in the following locations: Cell membrane, Cytoplasm, Early endosome, Nucleus, Peripheral membrane protein. Immunogen sequence: QFSYS
AKT2 is a phosphoinositide-dependent serine/threonine protein kinase that is enriched in insulin responsive tissues and has been implicated in the metabolic actions of the hormone. Akt is involved in many cellular signaling pathways and acts as a transducer of many functions initiated by growth factor receptors that activate phosphatidylinositol 3-kinase (PI 3-kinase). Akt2 is amplified and overexpressed in some human cancers.Spécification
Phospho-AKT2 (Ser474) | |
Polyclonal | |
Unconjugated | |
Akt2 | |
2410016A19Rik; AKT serine/threonine kinase 2; Akt2; AKT2 kinase; AW554154; HIHGHH; murine thymoma viral (v-akt) homolog-2; murine thymoma viral (v-akt) oncogene homolog 2; PKB; PKB beta; PKBB; PKBBETA; PRKBB; protein kinase Akt-2; Protein kinase B beta; protein kinase B, beta; putative v-akt murine thymoma viral oncoprotein 2; rac protein kinase beta; RAC protein kinase beta RAC-PK beta; RAC-BETA; RAC-beta serine/threonine protein kinase; RAC-beta serine/threonine-protein kinase; RAC-PK-beta; thymoma viral proto-oncogene 2; v-akt murine thymoma viral oncogene homolog 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11652, 208, 25233 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.59 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P31751, P47197, Q60823 | |
Akt2 | |
A synthetic phosphorylated peptide around S474 of human AKT2 (NP_001617.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |