Learn More
Proteintech Phantom Dye Violet 510 Viability Dye
Phantom Dye Violet 510 is a fixable viability dye which reacts with protein amine groups in both dead and live cells.
Marque: Proteintech PD00005-100TESTS
Code nomenclature Nacres: NA.82
Informations supplémentaires : Poids : 0.00200kg
Description
- Due to the compromised cell membrane in dead cells the dye can bind to many more proteins inside the cell, resulting in much higher fluorescence and easy differentiation of Live/Dead cells.
- Phantom Dye Violet 510 is a water soluble violet fluorescent dye excited by the Violet laser (405 nm), emitting at 510 nm, making it an excellent addition to your multicolor panel.
Spécification
Flow Cytometry | |
510 Emission | |
100 Tests |