Learn More
Schmidlin™ PG Plus FID Station H2 and Zero Air Generator
This dual gas generator produces a continuous supply of high purity hydrogen and zero air.
Marque: Schmidlin™ DB-FHG250-EU
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Increase efficiency and improve performance
- On-demand supply 24/7
- H2 flow rate and zero air up to 5 L/min.
- THC Pressure: 11 barg (160 psig)
- Permeation membrane drying system
- USB connectivity
- Easy to install with 'plug and play' functionality
- Ideal for all GC detector applications
Spécification
68 cm | |
15 cm | |
11 bar | |
69 cm | |
110/120 V at 60 Hz, 220/240 V at 50 Hz | |
115 W |
250 mL/min. | |
35°C | |
27 in. | |
Outlet: 1/8 in. | |
>99.9996% |