missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539334
This item is not returnable.
View return policy
Description
Peroxin 5 (Pex5) and Peroxin 7 (Pex7), are receptors for proteins that contain the peroxisome targeting signal 1 (PTS1) and 2 (PTS2), respectively, and shuttle these proteins from the cytosol to the peroxisome. Peroxin 5, also designated PTS1 receptor, is expressed as two isoforms, Pex5L and Pex5S. Pex5L transports PTS1 and Pex7-PTS2 cargo complexes to the initial Pex5 docking site, Pex14, while Pex5S transports only PTS1 cargoes. Pex5 and Pex7 also require either direct or indirect interaction with Peroxin 13 (Pex13) for proper import into peroxisomes. Mutations in the peroxin genes result in peroxisome biogenesis disorders (PBDs). Defects in the Pex5 gene are linked to Zellweger syndrome (cerebro-hapato-renal syndrome) of complementation group 2 (CG2), the most severe form of PBDs.
Specifications
PEX7 | |
Polyclonal | |
Unconjugated | |
PEX7 | |
PCDP1, Peroxin 7, Peroxisome biogenesis factor 7, PEX7, PEX7 protein, PTS2 receptor, PTS2R, RD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18634, 308718, 5191 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O00628, P97865 | |
PEX7 | |
A synthetic peptide derived from the internal region of human PEX7 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
PEX7 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction