Learn More
Sampling Systems™ Pendulum Beaker
Tous les produits Sampling Systems Produits
Click to view available options
Capacité (métrique):
250 ml
500 ml
1 l
2 l
Graduée:
Non
Oui
Matériau:
Acier inoxydable
Polypropylène
Conditionnement:
1 pièce
Description
- Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
- A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
- Attachments are interchangeable and simply click into position
Spécification
Spécification
| Type de produit | Swabbing Template |
| Matériau | Polypropylène |
| Autoclavable | Autoclavable |
| Capacité (métrique) | 1 l |
| Jetable | Reusable |
| Graduée | Non |
| Stérilité | Non stérile |