Learn More
DiscoverX PathHunter™ eXpress CCR1 Activated GPCR Internalization Assay
The PathHunter™ eXpress CCR1 Activated GPCR Internalization Assay measures CCR1 (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.
Marque:  DiscoverX 93-0651E3CP5M
Encompass
Code produit. 17333599
  • 2683.50€ / 1 kit
Expédition estimée: 28-03-2025
pour voir le stock disponible 
Découvrez plus d'offres spéciales
Ajouter au panier
Description
Description
The PathHunter™ CCR1 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. CCL3/MIP-1α) based internalization of CCR1 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged CCR1. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting CCR1-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Spécification
Spécification
GPCR
200 dp (2 x 96-well)
CCR1
Assay-Ready eXpress Kits
CCR1
Suggestions de produits
Suggestions de produits
Vidéos
FDS
Documents
Documents
One moment while we fetch your results.
Certificats
Promotions
Promotions
Correction du contenu d'un produit
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.
Nom du produit
DiscoverX PathHunter™ eXpress CCR1 Activated GPCR Internalization Assay