missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55149-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is specific to PARP2 (N-terminal).
This gene encodes polytransferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly transferase. The basic residues within the N-terminal region of this protein may bear potential DNA-binding properties, and may be involved in the nuclear and/or nucleolar targeting of the protein. Two alternatively spliced transcript variants encoding distinct isoforms have been found.Spécification
PARP2 | |
Polyclonal | |
Unconjugated | |
PARP2 | |
ADPRT 2, ADPRT2, ADPRTL2, ADPRTL3, hPARP 2, pADPRT 2, PARP 2, PARP2, poly (ADP ribose) polymerase 2, Poly [ADP ribose] polymerase 2, Poly[ADP ribose] synthase 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10038, 11546 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88554, Q9UGN5 | |
PARP2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |