Learn More
Kolmi Hopen™ Op-Air™ One Type IIR Splash-resistant Medical Mask with Foam
Splash-resistant medical mask with anti-fog nose piece to prevent glasses from fogging.
Marque: Kolmi Hopen™ M34101M-30
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Protects environment from droplets from wearer
- Provides protection against biological fluids
- Effective against bacteria, particles, and viruses
- Non-irritating inner layer
- Inner layer is specially designed to reduce moisture and wick away sweat
- Used by healthcare workers to reduce risk of infection or in the operating room
Spécification
Bleu | |
Polypropylène | |
CE, (UE) 2017/745, EN 14683:2019+AC:2019 | |
Médical |
Attaches | |
Plissée | |
Oui | |
40 Paquets |