Learn More
Thermo Scientific™ Nunc™ EasYFlask™ Flacons avec Nunclon™ Supra Surface
Code produit. p-200007435
Click to view available options
Surface de culture:
175 cm²
25 cm²
75 cm²
Type de bouchon:
Filtre
Plein
Conditionnement:
100 pièces
200 pièces
30 pièces
Code produit. 30408805
Marque: Thermo Scientific™ 156372
Les flacons Nunc EasYFlask de Thermo Scientific avec traitement de surface Nunclon Supra offrent des performances optimales pour la culture d'une grande variété de cellules adhérentes fastidieuses.
Experience optimal performance with Thermo Scientific Nunc EasYFlask flasks with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. The ergonomic design with angled neck and wide opening allows easy access to the growth surface by pipettes or cell scrapers. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.
The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.
Features of Nunc EasYFlask flasks with Nunclon Supra surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years
TRUSTED_SUSTAINABILITY
Spécification
Description Nunc EasYFlask, 25 cm²
Nbre par carton 200 Flasks
Nb par paquet 10 Flacons
Type de bouchon Filtre
Surface de culture 25 cm²
Pour un usage en laboratoire