missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NT5C3 293T Cell Overexpression Lysate (Denatured), Abnova
Marque: N/A H00051251-T01.100uL
Code nomenclature Nacres: NA.55
Informations supplémentaires : Poids : 0.00010kg
Description
Western Blotting
Spécification
Western Blot | |
BC015856 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-NT5C3 antibody (H00051251-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
NT5C3 293T Cell Transient Overexpression Lysate(Denatured) | |
31.57 | |
pCMV-NT5C3 full-length | |
100 μL | |
Human |