missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ NPC2 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Marque: Abnova™
Code nomenclature Nacres: NA.55
Informations supplémentaires : Poids : 0.00010kg
Spécification
Western Blot | |
NPC2 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 16.72kDa | |
Transient overexpression cell lysate was tested with anti-NPC2 antibody (Mfr. No. H00010577-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
NM_006432.3 | |
pCMV-NPC2 full-length | |
100 μL | |
Human |
Sécurité et traitement
Mention d'avertissement
- Attention