Learn More
Novus Biologicals™ Human Multi-tissue Tissue MicroArray (Cancer)
Code produit. p-5162466 Tous les produits Bio Techne Produits
Code produit. 18736853
Marque: Novus Biologicals™ NBP230262
Applications: Immunohistochemistry-Paraffin
  • Formalin fixed
  • Species: Human
  • Cross reactivity: Human
  • Tissue condition: Cancer
  • No. of samples: 59
  • No. of patients: 59
  • Core diameter: 2.0mm
  • Section thickness: 4μm
Recommended Storage
Store at 4°C. Do not freeze
Garantie
1 Year
TRUSTED_SUSTAINABILITY
This product is for research use only and is not approved for use in humans or in clinical diagnosis.