Learn More
NOMO2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14328-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| NOMO2 | |
| Polyclonal | |
| Unconjugated | |
| NOMO2 | |
| NODAL modulator 2, Nomo, NOMO2, PM5, pM5 protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 283820 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5JPE7 | |
| NOMO2 | |
| NOMO2 Fusion Protein Ag5595 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |