Learn More
Melford Nitrofurantoin
Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis.
40.10€ - 462.00€
Spécification
CAS 67-20-9
Formule moléculaire C8H6N4O5
Consulter d'autres spécifications
Produits 4
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
30018589
Afficher les documents
Melford
N51500-1.0
1 g
40.10€
1g
30018591
Afficher les documents
Melford
N51500-25.0
25 g
121.00€
25g
30018590
Afficher les documents
Melford
N51500-100.0
100 g
462.00€
100g
30018592
Afficher les documents
Melford
N51500-5.0
5 g
60.30€
5g
Description
Description
Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis.
Nitrofurantoin works by inhibiting bacterial enzymes involved in the metabolism of certain compounds, leading to the disruption of bacterial DNA, RNA, and protein synthesis.
Researchers may use nitrofurantoin as a selective agent by incorporating it into culture media to promote the growth of cells or organisms that have been genetically engineered to express resistance to this antibiotic. It may have applications in microbiology and cell culture experiments, particularly in the context of antibiotic selection.
Spécification
Spécification
67-20-9
C8H6N4O5