missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NINJ2 293T Cell Overexpression Lysate (Denatured), Abnova
Marque: N/A H00004815-T01.100uL
Code nomenclature Nacres: NA.55
Informations supplémentaires : Poids : 0.00010kg
Description
Western Blotting
Spécification
Western Blot | |
NINJ2 293T Cell Transient Overexpression Lysate(Denatured) | |
15.73 | |
pCMV-NINJ2 full-length | |
100 μL | |
Human |
293T | |
BC057766 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-NINJ2 antibody (H00004815-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Sécurité et traitement
Mention d'avertissement
- Attention