missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHP2L1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15802-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5′ stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.Spécification
NHP2L1 | |
Polyclonal | |
Unconjugated | |
SNU13 | |
15.5K, FA 1, FA1, hSNU13, NHP2 like protein 1, NHP2L1, NHPX, OTK27, SNRNP15 5, SNU13, SNU13 homolog, SPAG12, SSFA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20826, 4809 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P55769, Q9D0T1 | |
SNU13 | |
NHP2L1 Fusion Protein Ag8532 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |