missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
121.38€ - 325.38€
Spécification
Antigène | NFIB |
---|---|
Concentration | 0.18 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Description
NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.Spécification
NFIB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O00712, P97863 | |
18028, 29227, 4781 | |
NFIB Fusion Protein Ag10165 | |
Primary | |
-20°C | |
NFIB |
0.18 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CTF, HMGIC/NFIB, NF I/B, NF1 B, NFI B, NFI RED, NFIB, NFIB2, NFIB3, Nuclear factor 1 B type, Nuclear factor 1/B, nuclear factor I/B, TGGCA binding protein | |
Nfib | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
NFIB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech