Learn More
Neuropilin 2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marque: Invitrogen PA547762
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 25% cross&#8209;reactivity with recombinant human Neuropilin-2 is observed, and less than 5% cross-reactivity with recombinant rat Neuropilin-1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Neuropilin 2 Polyclonal antibody specifically detects Neuropilin 2 in Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Neutralization, Western Blot.Spécification
Neuropilin 2 | |
Polyclonal | |
Unconjugated | |
NRP2 | |
1110048P06Rik; neuropilin 2; neuropilin-2; neuropilin-2a(17); neuropilin-2a(22); neuropilin-2b(0); Np2; Np-2; Npn2; Npn-2; NRP2; PRO2714; receptor for VEGF165 and semaphorins class3; vascular endothelial cell growth factor 165 receptor 2; VEGF165R2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
18187, 81527 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O35276, O35375 | |
NRP2 | |
Mouse myeloma cell line NS0-derived recombinant rat Neuropilin-2 Gln23-Asp857 (Val809-Asp825 del) | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |