Learn More
Neuroligin 3/NLGN3 Antibody (242), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP2-89824H
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Neuroligin 3/NLGN3 Monoclonal antibody specifically detects Neuroligin 3/NLGN3 in Human samples. It is validated for Flow Cytometry
Spécification
| Neuroligin 3/NLGN3 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 242 | |
| Flow Cytometry | |
| ASPGX1, AUTSX1, Gliotactin homolog, KIAA1480HNL3, neuroligin 3, neuroligin-3, NL3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Neuroligin 3/NLGN3 (Uniprot#: Q9NZ94-2; Met1-Ser689). | |
| 0.1 mL | |
| Neuroscience | |
| 54413 | |
| Store at 4C in the dark. | |
| IgG |