missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuregulin-3/NRG3 Antibody (06), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-06610PE
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Neuregulin-3/NRG3 Monoclonal antibody specifically detects Neuregulin-3/NRG3 in Human samples. It is validated for ELISA
Spécification
| Neuregulin-3/NRG3 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 06 | |
| ELISA | |
| HRG3, neuregulin 3, neuregulin 3 variant 1, neuregulin 3 variant 10, neuregulin 3 variant 11, neuregulin 3 variant 12, neuregulin 3 variant 13, neuregulin 3 variant 14, neuregulin 3 variant 3, neuregulin 3 variant 4, neuregulin 3 variant 5, neuregulin 3 variant 6, neuregulin 3 variant 7, neuregulin 3 variant 8, neuregulin 3 variant 9, neuregulin-3-like polypeptide, pro-neuregulin-3, membrane-bound isoform, pro-NRG3 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Neuregulin-3/NRG3 (Accession#: ADK90021.1; Glu282-Pro342). | |
| 0.1 mL | |
| Cancer, Neuroscience, Signal Transduction | |
| 10718 | |
| Store at 4C in the dark. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu