Learn More
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Tubes
Description
Neon NxT Electroporation System 8-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 8-Channel Pipette (Cat. Nos. NEON18S, NEON18SK, NEON8, NEON8U). Each Neon NxT Electroporation System Kit with 8-Channel Tubes for 96 x 2 reactions (Cat. Nos. N1096-8 and N10096-8) comes with four 8-channel tubes and each Neon NxT Electroporation System Kit for 384 x 2 reactions (Cat. Nos. N10384-8 and N100384-8) comes with sixteen 8-channel tubes. This pack of eight tubes is offered for those wishing to use one tube per eight reactions (tubes can be used up to twelve times) depending on their experimental setup.
Spécification
Spécification
| Contenu et stockage | Includes 8 x 8-chamber tubes, use at room temperature under normal laboratory conditions. Refer to the Neon™ NxT Electroporation System user manual for protocol instructions. |
| Comprend | Includes 8 x 8-chamber tubes, use at room temperature under normal laboratory conditions. Refer to the Neon™ NxT Electroporation System user manual for protocol instructions. |
| Quantité | 8 Tubes,8 Tubes |
| Type de cellule | Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | Electroporation Pipette Tip |
| À utiliser avec (équipement) | Neon™ NxT Electroporation System |
| Transfection Technique | Electroporation |
| À utiliser avec (application) | Transfection |