missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV2 293T Cell Overexpression Lysate (Denatured), Abnova
Marque: N/A
Code nomenclature Nacres: NA.55
Informations supplémentaires : Poids : 0.00010kg
Description
Western Blotting
Spécification
Western Blot | |
NDUFV2 293T Cell Transient Overexpression Lysate(Denatured) | |
27.5 | |
pCMV-NDUFV2 full-length | |
100 μL | |
Human |
293T | |
NM_021074 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-NDUFV2 antibody (H00004729-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Sécurité et traitement
Mention d'avertissement
- Attention