missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA4L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16480-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NDUFA4L2 is a protein coding gene.Spécification
NDUFA4L2 | |
Polyclonal | |
Unconjugated | |
NDUFA4L2 | |
NDUFA4L2, NUOMS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100362331, 407790, 56901 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NRX3 | |
Ndufa4l2 | |
NDUFA4L2 Fusion Protein Ag9600 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |