Learn More
NDFIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 15602-1-AP-150UL
 Afficher plus de versions de ce produit
Code produit. 16844844
  • 410.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
The NEDD4 family-interacting protein 1 (NDFIP1) belongs to a small group of evolutionarily conserved proteins with three transmembrane domains and is an integral Golgi membrane protein. It is a potential target for ubiquitination by the Nedd4 family of proteins. NDFIP1 is strongly expressed in surviving neurons following acute cortical brain injury, and overexpression in cultured cortical neurons increased survival following growth factor starvation, suggesting that NDFIP1 may play a role in neuronal survival. NDFIP1 and the related protein NDFIP2 are thought to interact with and regulate multiple components of the EGF and PTEN/Akt signaling pathways. Recent studies suggest that NDFIP1 may also play a role in Th17 differentiation by limiting the production of proinflammatory cytokines.
Spécification
Spécification
NDFIP1
Polyclonal
Unconjugated
NDFIP1
N4WBP5, NDFIP1, PSEC0223
Rabbit
Antigen Affinity Chromatography
RUO
291609, 65113, 80762
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5U2S1, Q8R0W6, Q9BT67
Ndfip1
NDFIP1 Fusion Protein Ag7985
150 μL
Primary
Human, Mouse, Rat