Learn More
Tocris™ NBC 6
NLRP3 inflammasome inhibitor
Marque:  Tocris™ 6925/50
Code produit. 18737476
  • 942.74€ / 50mg
Découvrez plus d'offres spéciales
Description
Description
NBC 6 is a NLRP3 inflammasome inhibitor (IC50 = 574 nM). Inhibits nigericin-induced inflammasome activation in THP-1 cells and Imiquimod (Cat. No. 3700) induced IL-1β release from LPS-primed bone marrow-derived macrophages (BMDMs).
Spécification
Spécification
(T4)-[2-Acetyl-4,4,4-trichloro-3-(imino-κN)butanamidito-κO]diphenylboron
50 mg
C18H16BCl3N2O2
98%
2068818-02-8
Inflammasomes Inhibitors
7131