Learn More
Tocris™ NBC 6
NLRP3 inflammasome inhibitor
Marque: Tocris™ 6925/10
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
NBC 6 is a NLRP3 inflammasome inhibitor (IC50 = 574 nM). Inhibits nigericin-induced inflammasome activation in THP-1 cells and Imiquimod (Cat. No. 3700) induced IL-1β release from LPS-primed bone marrow-derived macrophages (BMDMs).Spécification
(T4)-[2-Acetyl-4,4,4-trichloro-3-(imino-κN)butanamidito-κO]diphenylboron | |
10 mg | |
C18H16BCl3N2O2 | |
98% |
2068818-02-8 | |
Inflammasomes Inhibitors | |
1869 |