missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natamycin, MedChemExpress
Click to view available options
Quantité:
50 mg
100 mg
200 mg
500 mg
Description
Natamycin (Pimaricin) is a macrolide antibiotic agent produced by several Streptomyces strains. Natamycin inhibits the growth of fungi via inhibition of amino acid and glucose transport across the plasma membrane. Natamycin is a food preservative, an antifungal agent in agriculture, and is widely used for fungal keratitis research.
Spécification
Spécification
| Nom chimique ou matériau | Natamycin |
| Forme physique | Solid |
| CAS | 7681-93-8 |
| Couleur | Jaune clair |
| Quantité | 500 mg |
| Notes de qualité de la pureté | Research |
| Formule moléculaire | C33H47NO13 |
| À utiliser avec (application) | COVID-19-immunoregulation |
| Synonyme | Pimaricin |
| Informations sur la solubilité | DMSO : 16.67 mg/mL (25.04 mM; ultrasonic and adjust pH to 6 with HCl) ∣H2O : 1 mg/mL (1.50 mM; ultrasonic and warming and heat to 80°C) |
| Afficher plus |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction