Learn More
N-Boc-L-isoleucinol, 95 %, Thermo Scientific Chemicals
Code produit. 11324276
Click to view available options
Quantité:
1 g
5 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 106946-74-1
Formule moléculaire C11H23NO3
Poids moléculaire (g/mol) 217.31
Numéro MDL MFCD00235929
Clé InChI BPLDQMXXYMKQPW-DTWKUNHWSA-N
Synonyme n-boc-l-isoleucinol, boc-isoleucinol, n-boc-l-isolucinole, n-boc-2s,3s---2-amino-3-methyl-1-pentanol, boc-l-isoleucinol, tert-butyl 2s,3s-1-hydroxy-3-methylpentan-2-yl carbamate, boc-ile-ol, ambotzbal1038, pubchem15801
CID PubChem 14237729
Nom IUPAC N-[(2S,3S)-1-hydroxy-3-méthylpentan-2-yl]carbamate de tert-butyle
SMILES CC[C@H](C)[C@@H](CO)NC(=O)OC(C)(C)C
Spécification
Nom chimique ou matériau N-Boc-L-isoleucinol
Point d’ébullition 121°C to 126°C (0.6mmHg)
Indice de réfraction 1.453
Beilstein 3604099
Quantité 1 g
Poids de la formule 217.31
Pourcentage de pureté 95%