Learn More
MYCT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 22004-1-AP-20UL
Code produit. 16851545
  • 145.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
MYCT1 was initially identified as a novel target of the c-Myc oncogene in myeloid cells. It is a widely expressed nuclear protein whose overexpression can promote apoptosis, alteration of morphology, enhancement of anchorage-independent cell growth, tumorigenic conversion, promotion of genomic instability, and inhibition of hematopoietic differentiation. MYCT1 binds to the promoters of several c-Myc-regulated genes and it has been suggested that the phenotypes seen in MYCT1-overexpressing cells are a result of the deregulation of these genes. RUNX1-ETO, a fusion protein made up of RUNX1 and ETO, is thought to deregulate the proliferation and responsiveness of human hematopoietic progenitor cells downstream of MYCT1.
Spécification
Spécification
MYCT1
Polyclonal
Unconjugated
MYCT1
MTLC, MTMC1, myc target 1, Myc target protein 1, MYCT1
Rabbit
Antigen Affinity Chromatography
RUO
68632, 80177
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N699, Q8R411
Myct1
MYCT1 Fusion Protein Ag4800
20 μL
Primary
Human, Mouse