missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benchmark Scientific™ myBlock HL Mini Digital Dry Bath with Heated Lid
Click to view available options
myBlock HL Mini Digital Dry Bath with Heated Lid; Holds: 0.2, 0.5, and 1.5 mL Microtubes
Description
- Ideal for temperature sensitive applications including immunoassays, LAMP assays and enzyme reactions
- Independent temperature control of lid and block
- Compact, fits in the palm of your hand
- Room temperature to 37°C in just 2 min.
- Exchangeable blocks for 0.2 to 1.5 mL tubes
- Blocks sold separately
Spécification
Autoclavable | Not Autoclavable |
Capacité (métrique) | 40 x 0.2 mL, 24 x 0.5 mL, 15 x 1.5 mL |
Matériau de la cuve | Aluminum |
Technologie de capteur de CO2 | No Sensor |
À utiliser avec (équipement) | Microtubes |
Capacité | 0.2, 0.5, and 1.5 mL Microtubes |
Éclairage | No Lighting |
Matériau | Plastic |
No. of Places | 1 |
Contrôle de l’oxygène | None |
Afficher plus |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'