missing translation for 'onlineSavingsMsg'
Learn More

MYBBP1A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Spécification

Antigène MYBBP1A
Concentration 0.35 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation
Classification Polyclonal
Conjugué Unconjugated
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
16833204
Afficher les documents
Proteintech
14524-1-AP-20UL
20 μL
145.00€
20µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16823204
Afficher les documents
Proteintech
14524-1-AP-150UL
150 μL
410.00€
150µL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

The Myb family of transcription factors, which includes the structurally related A-, B-, and c-Myb genes, regulate differentiation, cellular growth and transactivating gene expression. c-Myb plays an essential role in controlling the proliferation and differentiation of hematopoietic cells, cellular levels of c-Myb decreasing as cells reach terminal differentiation. Myb-binding protein 1A (MYBBP1A) is a novel nuclear protein localized predomitly, but not exclusively, in nucleoli. Although initially isolated as a c-Myb-interacting protein, MYBBP1A is expressed ubiquitously, and associates with a number of different transcription factors. MYBBP1A associates with the aromatic hydrocarbon receptor (AhR), enhancing transactivation and increasing AhR-dependent gene expression. MYBBP1A is a powerful negative regulator of PPAR gamma coactivator 1 alpha (PGC-1 alpha) by p38 MAPK and also acts as a nucleocytoplasmic shuttling protein that utilizes CRM1-dependent and independent nuclear export pathways.
Spécification

Spécification

MYBBP1A
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation
Unconjugated
Rabbit
Human
Q9BQG0
10514
MYBBP1A Fusion Protein Ag6008
Primary
-20°C
MYBBP1A
0.35 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
MYB binding protein (P160) 1a, Myb binding protein 1A, MYBBP1A, P160, PAP2
MYBBP1A
IgG
Antigen Affinity Chromatography
Antibody
Vidéos
FDS
Documents

Documents

Product Certifications