Learn More
Manutan MW20G Multifunction Microwave
With an adjustable thermostat from 60° to 200°C.
Marque: Manutan A104510
Code nomenclature Nacres: NB.76
Informations supplémentaires : Poids : 11.72000kg
Description
- Oven capacity of 20L
- Electromechanical programming
- Five functions: microwave only, defrost, favorite preparation, single grill, combined grill mode
- Glass turntable
- Timer to 60 min, 24-hour clock
- The grid at the top makes your dishes crispy on the outside yet tender inside
Spécification
20 L | |
24.5 cm Diameter Glass Turntable | |
33.5 cm | |
24.5 cm | |
11 kg | |
1050 W |
Multi-Purpose Oven | |
MW20G | |
44 cm | |
33.5 cm | |
230 V |