Learn More
Tocris Bioscience™ MRS 2500 tetraammonium salt
Highly potent and selective P2Y1 antagonist
Marque: Tocris Bioscience™ 2159/1
Code nomenclature Nacres: NA.22
Informations supplémentaires : Poids : 0.00970kg
Description
MRS 2500 tetraammonium salt is a highly potent and selective antagonist of the platelet P2Y1 receptor (Ki = 0.78 nM). Inhibits ADP-induced aggregation of human platelets with an IC50 value of 0.95 nM. Inhibits the upregulation of NTPDase1 by ATPγS. Prevents thrombus formation in vivo.Spécification
>98% | |
MRS 2500 tetraammonium salt | |
MRS 2500 tetraammonium salt |
1 mg | |
629.29 |
For Research Use Only