Learn More
Tocris Bioscience™ MRS 2500 tetraammonium salt
Highly potent and selective P2Y1 antagonist
Marque:  Tocris Bioscience™ 2159/1
Code nomenclature Nacres: NA.22
Informations supplémentaires : Poids : 0.00970kg
Code produit. 18065982
  • 481.40€ / 1mg
Expédition estimée: 18-07-2024
pour voir le stock disponible 
Description
Description
MRS 2500 tetraammonium salt is a highly potent and selective antagonist of the platelet P2Y1 receptor (Ki = 0.78 nM). Inhibits ADP-induced aggregation of human platelets with an IC50 value of 0.95 nM. Inhibits the upregulation of NTPDase1 by ATPγS. Prevents thrombus formation in vivo.
Spécification
Spécification
>98%
MRS 2500 tetraammonium salt
MRS 2500 tetraammonium salt
1 mg
629.29