missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse XCL1/Lymphotactin Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Marque: R&D Systems AF486
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
XCL1/Lymphotactin Polyclonal specifically detects XCL1/Lymphotactin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Spécification
| XCL1/Lymphotactin | |
| Polyclonal | |
| Unconjugated | |
| ATAC, ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 alpha, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6375 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 5-15 ug/mL | |
| XCL1 | |
| E. coli-derived recombinant mouse XCL1/Lymphotactin | |
| 100 μg | |
| Primary | |
| Detects XCL1/Lymphotactin in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) GRO alpha , rhGRO beta , rhGRO gamma , and recombinant mouse MCP-5 is observed. | |
| Mouse | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu